Acetonitrile : water : formic acid (5:95:0.1)%, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # BJLC583-4
Supplier: Honeywell Research Chemicals
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:BJLC583-4
- Supplier no.:LC583-4
Specifications
About this item
:
Boiling Pt:82 °C (solvent)
Density:0.9927 g/cm³ (20 °C)
Flash Pt:43.3 °C
:GHS08
:GHS02
Storage Temperature:Ambient
:64-18-6