Dichloromethane ≥99.9% stabilized, B&J Brand™ for applications requiring low water content, Burdick & Jackson™
Catalog # BJCS301-200
Supplier: Honeywell Research Chemicals
Product available on GSA Advantage®
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:200 L
- Cat. No.:BJCS301-200
- Supplier no.:CS301-200
Specifications
Specification Test Results
- Acidity (as HCl):1 mg/L max.
- Chloride:10 mg/L max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.4236 - 1.4246
- Residue:1 mg/L max.
- UV Absorbance @ 240 nm:0.1 AU max.
- UV Absorbance @ 250 nm:0.01 AU max.
- UV Cutoff:233 nm max.
- Water by Karl Fischer Titration:0.003 % max.
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1.33 g/cm³
Flash Pt: Does not flash
EINECS:200-838-9
UN:1593
ADR:6.1,III
:Stabilised with cyclohexene
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881