U-Tube Viscometer Type BS/U Capillary Viscometers, Fungilab
Supplier: Avantor
Certificates
About this item
BS U Tube Reverse Flow capillary viscosmeter for opaque liquids
Made of borosilicate glass, and non-alterable timing mark.
Certifications: BS, UNE 400313, ASTM D-445, D446, ISO 3104, ISO 3105.
Ordering information: Includes constant value calibration certificate at 40ºC (104ºF) and 100ºC (212ºF).
Frequently Bought Together