About this item
Not only do anaerobic microorganisms require the environment to be oxygen free, but a reduced environment is physiologically necessary for anaerobes to produce energy for growth
The Oxyrase® Enzyme System used in OxyPRAS Plus® plates provides a reduced medium before sterilization and maintains the medium in a reduce state for storage and during use. The Oxyrase® Enzyme System prevents the formation of undesirable oxidation products in these PRAS plates. Growth of anaerobes on OxyPRAS Plus® plates require anaerobic incubation in jars, bags, or chambers.
Types of OxyPRAS Plus® Plates Available, and Their Uses: Brucella Blood Agar (PBRW): general media for variety of bacteria. AnaSelect® Blood Agar (PBRW/AS): designed for mixed cultures. Kanamycin / Vancomycin Laked Blood Agar (PCDL): inhibitory. Bile Bacteroides Esculin (PBBN): differentiation. Bacteroides sp.. BBE / KVL Bi-plate (BBCDKV): used as a dual purpose plate. Phenylethyl Alcohol Blood Agar (PPEW): mixed cultures (G+/G-).
Sterile plates, aseptically packed individually, and ready for immediate use, with jars, bags, and or chambers.
Each OxyPRAS Plus® plate conforms to the specifications of PRAS media, and are used for the isolation and cultivation of anaerobic bacteria from a variety of clinical and non-clinical materials. It can be stored in the range of 20°C to 25°C, if used at RT.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Storage Conditions:2 - 8 °C