ContactUsLinkComponent
OxyPouch™ Oxygen Barrier Bag, Oxyrase®
OxyPouch™ Oxygen Barrier Bag, Oxyrase®
Catalog # 101975-878
Supplier:  Oxyrase Inc
OxyPouch™ Oxygen Barrier Bag, Oxyrase®
Catalog # 101975-878
Supplier:  Oxyrase Inc
Supplier Number:  A-OP-0100
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    100 individual bags
  • Cat. No.:
    101975-878
  • Supplier no.:
    A-OP-0100
Specifications
About this item
The OxyPouch™ is a foil oxygen barrier bag, designed and specifically sized to hold OxyPlates (or other type petri dishes) and help prevent oxidation of plates during incubation and or storage.
The OxyPouch™ is designed and constructed of materials to ensure that once the bag has been properly sealed with a heat sealer, oxygen cannot permeate through the bag. This prevents any further oxidation of the plate sealed within the bag.
Each OxyPouch™ also has one clear side with which to easily read the labeled portion of the base plate, without the need to open the bag. The upper portion of the bag is perforated, to make opening simple, without the need for scissors.
Ordering information: OxyPouch™ is shipped direct to customers.