NxGen™ Rnase Inhibitor
Supplier: Lucigen
Certificates
About this item
Eliminate RNAse activity with Lucigen’s NxGen™ RNAse Inhibitor.
RNAse Inhibitor is a potent inhibitor of ribonucleases such as RNase A, RNase B, and RNase C. The 52 kDa protein is a fusion of the porcine RNAse Inhibitor gene with a proprietary 22.5 kDa protein tag.
Performance Specifications: 1) Unit Concentration: 40,000 U/mL. 2) Purity (SDS-PAGE): >99%. 3) SS Exonuclease: 2000 U <5.0% released. 4) DS Exonuclease: 2000 U <0.5% released. 5) Endonuclease: 2000 U <10% converted. 6) Non-specific RNAse: 2000 U none detected. 7) E. coli 16S rDNA Contamination: 2000 U <10 copies. 8) Storage: -20°C.
Source: A recombinant E. coli strain carrying the porcine RNAse Inhibitor gene.
Ordering information: Supplied at 40,000 U/mL in 20 mM HEPES-KOH, 50 mM KCl, 8 mM dithiothreitol, 50% glycerol, pH 7.5 @ 25°C. Unit definition: One unit is defined as the amount of enzyme required to inhibit by 50% the hydrolysis of cytidine 2',3'-cyclic monophosphate by 5 ng of RNAse A.
Specifications
- Protein/Peptide Name:Rnase Inhibitor