Order Entry
United States
ContactUsLinkComponent
OxyFluor™ Fluorescence Microscopy, Oxyrase®
OxyFluor™ Fluorescence Microscopy, Oxyrase®
Catalog # 101975-866
Supplier:  Oxyrase Inc
undefined
OxyFluor™ Fluorescence Microscopy, Oxyrase®
Catalog # 101975-866
Supplier:  Oxyrase Inc
Supplier Number:  OF-0005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Bottle, 5 ml
  • Cat. No.:
    101975-866
  • Supplier no.:
    OF-0005

Specifications

About this item

OxyFluor™ (OF) is an enzyme formulation that has been designed for use in fluorescence microscopy to prevent photo-bleaching and photo-damage by removing dissolved oxygen directly into water, at the request of researchers in the field

The formulation of OxyFluor™ combines Oxyrase®, and DL-Lactate (100mM) to stabilize the Oxyrase® in suspension. Depending upon conditions of use, 10 to 20mM of DL-Lactate and/or DL-succinate subtrates may need to be added to ensure efficiency.

One unit of OxyFluor™ activity will reduce dissolved oxygen (in 1mL of air saturated 40mM phosphate Buffer, pH 8.4, at 37C) at the rate of 1% per second, and the rate of oxygen removal increases with temperature. However, above 55°C, it is rapidly inactivated. OxyFluor™ is active over a wide pH range of 6.8 to 8.4. It can be stored in the range of 2° to 8°C, if used within 30 days.

OF also continues to keep oxygen out of the specimen being observed over extended periods of time. OxyFluor™ is non-sterile, but has a bioburden less than 103 cfu per milliliter.

Ordering information: OxyFluor™ has a shelf life of 18 months under recommended storage and use conditions. Oxyrase® Inc. guarantees a minimum of 6 months shelf life from shipment date. If a longer shelf life is needed, this should be arranged at the time the order is placed (OxyFluor™ is shipped direct to customer.).