Corning® TE Buffer, 1X, Liquid, Corning
Catalog # 45001-155
Supplier: Corning
CAS Number:
Specifications
- Description:1X TE Buffer, Liquid
- Size:6×1 L
- Storage Temperature:15...30 °C
- Cat. No.:45001-155
- pH:7.9 - 8.1
- Supplier no.:46-009-CM
Specifications
About this item
Tris-EDTA (TE) buffer is commonly used in work with nucleic acids.
Examples include as a solubilization buffer in DNA and RNA purification, as a running buffer in electrophoresis, and as a general storage buffer for nucleic acids.