Order Entry
United States
ContactUsLinkComponent
Antimony ICP-MS Standard, 1 ml = 1 mg Sb (1000 ppm Sb)Sb in H₂O / Tartaric Acid / trace HNO₃, Ricca Chemical Company
Antimony ICP-MS Standard, 1 ml = 1 mg Sb (1000 ppm Sb)Sb in H₂O / Tartaric Acid / trace HNO₃, Ricca Chemical Company
Catalog # RCMSSB1KW-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Antimony ICP-MS Standard, 1 ml = 1 mg Sb (1000 ppm Sb)Sb in H₂O / Tartaric Acid / trace HNO₃, Ricca Chemical Company
Catalog # RCMSSB1KW-100
Supplier:  Ricca Chemical
Supplier Number:  RMSSB1KW-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O/Tartaric acid/tr. HNO₃
  • Size:
    100 ml
  • Cat. No.:
    RCMSSB1KW-100
  • Element:
    Antimony
  • Supplier no.:
    RMSSB1KW-100N

Specifications

About this item