Magnesium nitrate (2.0% Mg) in nitric acid 5% matrix modifier solution
Catalog # RCMMMG20KN-50
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:50 ml
- Cat. No.:RCMMMG20KN-50
- Supplier no.:RMMG20KN-50N
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless
- Certified Concentration:20,000 ± 1000 ppm Mg
Specification Test Results
About this item
:
UN:1474
ADR:5.1,III
CAS Number:10377-60-3
:13446-18-9
Formula:Mg(NO₃)₂