Order Entry
United States
Orders LinkContactUsLinkComponent
 

Trichloroacetic acid (TCA) 33% (w/v) in aqueous solution

 
 
 
 
Trichloroacetic acid (TCA) 33% (w/v) in aqueous solution
 
Trichloroacetic acid (TCA) 33% (w/v) in aqueous solution
Trichloroacetic acid (TCA) 33% (w/v) in aqueous solution

 

 
   
 
 :  TCA, Trichloroethanoic acid, Aceto-Caustin, TCA
  •  :76-03-9
  •  :Cl₃CCOOH
  •  :MFCD00004177
  •  :163.39 g/mol
  •  :~ 100 °C
  •  :~ 0 °C
  •  :1.1541 g/cm³
  •  :Ambient
  •  :2564
  •  :8,III
 

 

  • Appearance:
    Clear, colorless, mild Trichloroacetic odor
  • Assay at 20 °C (traceable to NIST SRM 84):
    33.0 ± 0.4 % (w/v)
Product Family Options

 

  • Pack type
    Size
     
     
  • Glass bottle
    120 ml
       
       
    Each (120ml)
    Each (120ml) - $135.97
    $135.97
    Each (120ml)
       $135.97

     : R8688300-120C
  • Glass bottle
    500 ml
       
       
    Each (500ml)
    Each (500ml) - $110.24
    $110.24
    Each (500ml)
       $110.24

     : R8688300-500C
  • Glass bottle
    1 L
       
       
    Each (1l)
    Each (1l) - $196.55
    $196.55
    Each (1l)
       $196.55

     : R8688300-1C

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.