

Sulkowitch reagent for qualitative detection of calcium in urine
Catalog # RC836016
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC836016
- Supplier no.:R8360000-500A
Specifications
Specification Test Results
- Appearance:Clear, colorless, Acetic odor
Specification Test Results
About this item
:
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0300 g/cm³
:GHS08
:GHS05
Storage Temperature:Ambient
:6009-70-7, 6153-56-6