
5-Sulfosalicylic acid 10% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:5965-83-3
:
Formula:HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MDL Number:MFCD00007508
:254.22 g/mol
Boiling Pt:~ 100 °C
Density:1.0600 g/cm³
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, slight tannish gold to slight pink, odorless
- Assay at 20 °C (traceable to NIST SRM 84):10.00 ± 0.02 % (w/v)
Specification Test Results
Related Information
Frequently Bought Together