
Sodium sulfite 10% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7757-83-7
:
Formula:Na₂SO₃
MDL Number:MFCD00003503
:126.04 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.1000 g/cm³
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 136):10.00 ± 0.06 % (w/v)
Specification Test Results
Frequently Bought Together