Silver nitrate 0.0100 N (N/100) in denatured alcohol
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
UN:1493
ADR:5.1,II
Specification Test Results
- Appearance:Clear, colorless, alcohol odor
- Assay at 20 °C (traceable to NIST SRM 999):0.01000 ± 0.00005 N at 20.0 °C
Specification Test Results
Related Information
Frequently Bought Together