
Ammonium thiocyanate 8% (w/v) in aqueous solution USP Test Solution (TS)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1762-95-4
:
Formula:NH₄SCN
MDL Number:MFCD00011428
:76.12 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0150 g/cm³
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):8.00 ± 0.05 % (w/v)
Specification Test Results
Frequently Bought Together