
Diammonium hydrogen orthophosphate solution 400 g/800 mL in water
Catalog # RC69016
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC69016
- Supplier no.:R0690000-500A
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless to slight ammonia odor
Specification Test Results
About this item
CAS Number:7783-28-0
:
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891
:132.06 g/mol
Density:1.2000 g/cm³
:GHS07
Storage Temperature:Ambient