Diammonium hydrogen orthophosphate 30% (w/v) in aqueous solution (1 ml = 1 mg Phenol) APHA for magnesium
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7783-28-0
:
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891
:132.06 g/mol
Density:1.2000 g/cm³
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, colorless liquid
Specification Test Results
Frequently Bought Together