Potassium Standard, 1 ml = 0.100 mg K, 100 ppm K KNO₃ in 3% HNO₃, RICCA Chemical Company
Catalog # RC6498-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:3% HNO₃
- Size:100 ml
- Cat. No.:RC6498-100
- Element:Potassium
- Supplier no.:R6498000-100A