Potassium permanganate 4% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7722-64-7
:
Formula:KMnO₄
MDL Number:MFCD00011364
:158.03 g/mol
UN:1490
ADR:5.1,II
Specification Test Results
- Appearance:Clear, dark purple, odorless
- Assay:4.00 ± 0.08 % w/v
Specification Test Results
Frequently Bought Together