

Ammonium chloride 8,5% (w/v) aqueous solution, with Brij®35
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:12125-02-9
:
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0255 g/cm³
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):8.50 ± 0.05 % (w/v)
Specification Test Results
Frequently Bought Together