
Ammonium carbonate solution R 158 g/L
Catalog # RC622.15-16
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC622.15-16
- Supplier no.:R0622150-500A
Specifications
Specification Test Results
- Appearance:Clear, colorless, ammonia odor
- Assay at 20 °C (traceable to NIST SRM 723):158.0 ± 0.8 g/L
Specification Test Results
About this item
:
:96.086 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0500 g/cm³
:GHS07
Storage Temperature:Ambient
:506-87-6
Formula:CH₈N₂O₃