ContactUsLinkComponent
Potassium Chloride Conductivity Standard, 2.767 mS/cm (2767 µmho/cm) at 25 °C, 0.02 Molar, Ricca Chemical Company
Potassium Chloride Conductivity Standard, 2.767 mS/cm (2767 µmho/cm) at 25 °C, 0.02 Molar, Ricca Chemical Company
Supplier:  Ricca Chemical
Certificates
undefined
Potassium Chloride Conductivity Standard, 2.767 mS/cm (2767 µmho/cm) at 25 °C, 0.02 Molar, Ricca Chemical Company
Supplier:  Ricca Chemical
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.