


Platinizing solution
Catalog # RC58624
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:120 ml
- Cat. No.:RC58624
- Supplier no.:R5862000-120C
Specifications
Specification Test Results
- Appearance:Clear, orange color, odorless
Specification Test Results
About this item
:
:96.7 g/mol
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0000 g/cm³
:GHS05
:GHS08
:GHS06
Storage Temperature:Ambient
:18497-13-7
Formula:PtCl₄·₂HCl·6H₂O