Order Entry
United States
ContactUsLinkComponent
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Catalog # RC5457-4
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Catalog # RC5457-4
Supplier:  Ricca Chemical
Supplier Number:  R5457000-120A
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    100 ppm
  • Size:
    120 ml
  • Cat. No.:
    RC5457-4
  • Contents:
    KNO₃ in H₂O/tr. CHCl₃
  • Supplier no.:
    R5457000-120A

Specifications

About this item