Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Ammonia (122 ppm NH₃), Ricca Chemical Company
Catalog # RC5453-4
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:122/100 ppm NH₃/N
- Pack type:Plastic Bottle
- Size:120 ml
- Cat. No.:RC5453-4
- Contents:NH₄Cl in H₂O
- Supplier no.:R5453000-120A