Nitrite Standard, 1 ml = 0.200 mg NO₂⁻, 200 ppm NO₂⁻ (60.9 ppm N), Ricca Chemical Company
Catalog # RC5444.2-4
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:200 ppm
- Size:120 ml
- Cat. No.:RC5444.2-4
- Contents:NaNO₂ in H₂O
- Supplier no.:R5444200-120C