Murexide 0.2% (w/w) in sodium chloride indicator
Catalog # RC5220-500
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:500 ml
- Cat. No.:RC5220-500
- Supplier no.:R5220000-500A5
Specifications
Specification Test Results
- Appearance:Rosy pink powder
- Suitability for Use:Pink (Excess Calcium) - Purple (Excess EDTA)
Specification Test Results
About this item
CAS Number:3051-09-0
:
Formula:C₈H₈N₆O₆
:284.19 g/mol
Boiling Pt:1461 °C
Melting Pt:801 °C
Density:2.1000 g/cm³
Storage Temperature:Ambient
MDL Number:MFCD00012777