Methyl orange 0.2% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:547-58-0
:
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502
:327.34 g/mol
UN:3143
ADR:6.1,III
Specification Test Results
- Appearance:Red, odorless
- Suitability for Use:Pink (pH 4.4)
Specification Test Results
Frequently Bought Together