Hydroxylammonium chloride 30% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:5470-11-1
:
Formula:NH₂OH·HCl
MDL Number:MFCD00051089
:69.49 g/mol
UN:2923
ADR:8,III
Specification Test Results
- Appearance:Clear, colorless, odorless
- Refractive Index:Confirmed concentration
Specification Test Results
Related Information
Frequently Bought Together