
Hydroxylamine - EDTA solution
Supplier: Ricca Chemical
Certificates
About this item
:7732-18-5
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0200 g/cm³
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, colorless, odorless
Specification Test Results
Frequently Bought Together