Crystal violet 2% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:548-62-9
:
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750
:407.99 g/mol
UN:3077
ADR:9,III
Specification Test Results
- Absorbance (traceable to NIST SRM 930):Conforms to control
- Appearance:Clear, dark violet, odorless
Specification Test Results
Frequently Bought Together