Order Entry
United States
ContactUsLinkComponent
Eosin Y (yellowish) working solution 0.25% (w/v) in 57% (v/v) alcohol, acidified stain
Health
Irritant
Flammable
Eosin Y (yellowish) working solution 0.25% (w/v) in 57% (v/v) alcohol, acidified stain
Supplier:  Ricca Chemical
Certificates
undefined
Health
Irritant
Flammable
Eosin Y (yellowish) working solution 0.25% (w/v) in 57% (v/v) alcohol, acidified stain
Supplier:  Ricca Chemical
Certificates

Specification Test Results

  • Absorbance (traceable to NIST SRM 930):
    Conforms to control
  • Appearance:
    Orange reddish, methanol odor
  • pH at 25 °C (traceable to NIST SRM 185 & 186):
    4.65 ± 0.02 pH at 25.0 °C
  • Specific Gravity:
    0.902 to 0.915 at 25°C

Specification Test Results

Related Information

Frequently Bought Together