Order Entry
United States
ContactUsLinkComponent
Cupri-citric solution R
Health
Corrosive
Cupri-citric solution R
Supplier:  Ricca Chemical
Certificates
undefined
Health
Corrosive
Cupri-citric solution R
Supplier:  Ricca Chemical
Certificates

Specification Test Results

  • Appearance:
    Clear, deep blue, odorless
  • Refractive Index:
    Concentration confirmed

Specification Test Results

Related Information

Frequently Bought Together