ContactUsLinkComponent
Congo red USP Test Solution (TS)
Health
Congo red USP Test Solution (TS)
Supplier:  Ricca Chemical
Certificates
undefined
Health
Congo red USP Test Solution (TS)
Supplier:  Ricca Chemical
Certificates
Specification Test Results
  • Appearance:
    Clear, red, acohol odor
  • Suitability for Use:
    Red (pH 5.0) - Blue (pH 3.0)