Order Entry
United States
ContactUsLinkComponent
Calcium IC Standard, 1 ml = 0.100 mg Ca, 100 ppm Ca CaCO₃in 3% HNO₃, Ricca Chemical Company
Calcium IC Standard, 1 ml = 0.100 mg Ca, 100 ppm Ca CaCO₃in 3% HNO₃, Ricca Chemical Company
Catalog # RC1795-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Calcium IC Standard, 1 ml = 0.100 mg Ca, 100 ppm Ca CaCO₃in 3% HNO₃, Ricca Chemical Company
Catalog # RC1795-100
Supplier:  Ricca Chemical
Supplier Number:  R1795000-100A
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    3% HNO₃
  • Size:
    100 ml
  • Cat. No.:
    RC1795-100
  • Element:
    Calcium
  • Supplier no.:
    R1795000-100A

Specifications

About this item