


Bromothymol blue 0.1% (w/v) in 50% (v/v) alcohol USP Test Solution (TS)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:76-59-5
:
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872
Boiling Pt:~ 82 °C
Density:0.9000 g/cm³
Flash Pt:24 °C
UN:1987
ADR:3,III
:624.39 g/mol
:GHS08
:GHS07
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, reddish orange, ethanol odor
- Suitability for Use:Yellow (pH 7.6)
Specification Test Results
Related Information
Frequently Bought Together