Boron Standard, 1 ml = 0.001 mg B, 1 ppm B, Ricca Chemical Company
Catalog # RC109016
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1 ppm
- Matrix:H₂O
- Size:500 ml
- Cat. No.:RC109016
- Element:Boron
- Supplier no.:R1090000-500A