Orders LinkContactUsLinkComponent
Upchurch Scientific® Fittings Kits, IDEX Health and Science
Upchurch Scientific® Fittings Kits, IDEX Health and Science
Supplier:   Upchurch Scientific
Upchurch Scientific® Fittings Kits, IDEX Health and Science
Certificates
Upchurch Scientific® Fittings Kits, IDEX Health and Science
Upchurch Scientific® Fittings Kits, IDEX Health and Science
7 Options Available Below
Product Details & Documents
MicroTight™ Tubing Sleeves facilitate capillary tubing connections with our MicroTight accessories
Manufacturer Specific Fittings Kits are customized for specific manufacturer’s legacy HPLC systems.
IDEX Health & Science manufactures a large selection of kits, each containing an assortment of products designed to make your life easier and to save you money. Our kits range from our Super Kit – which contains a collection of many products you’ll use to accessorize your systems – to more specialized kits like our FIA kit, offering products designed to help you with flow injection analyses.
Sleeves feature an outer diameter of .025" and are available in variety of inner diameters for a broad-range compatibility with a wide assortment of capillary tubing sizes. Manufactured from PEEK polymer, these sleeves carry an upper temperature threshold of 125 °C. For proper sleeve use, choose a sleeve with an inner diameter .001"-.002" (25-50 µm) larger than the outer diameter of your capillary tubing. Then slip the sleeve over the flow path tubing, such that the tubing extends all the way through the sleeve, but not beyond the end of the sleeve. Choose the correct fitting that corresponds with your receiving port, slide it over the sleeved flow path tubing and connect as normal.
Certifications: ROHS Certified
Product Family Options
Product Information
  • Material
    Color
    Tubing int.Ø
    Availability
    Price
  • 53498-532
    Kit, Fittings, Varian™ Compatible
    Kit, Fittings, Varian™ Compatible
    Multiple/Other
    Assorted
    N/A
    Discontinued
    Supplier Number: -
  • 53500-610
    Kit, MicroTight Connectors
    Kit, MicroTight Connectors
    PEEK
    Assorted
    Various
    Log in for availability
    Log in for availability
    Each
    Each - $380.24
    $380.24
    Each
       $380.24
    Supplier Number: 1356
  • 21001-110
    Kit, Flanged Fittings
    Kit, Flanged Fittings
    Multiple/Other
    Assorted
    N/A
    Log in for availability
    Log in for availability
    Each
    Each - $453.55
    $453.55
    Each
       $453.55
    Supplier Number: 1453
  • 21001-116
    Kit, Fittings, Low Pressure
    Kit, Fittings, Low Pressure
    ETFE, PEEK
    Assorted
    N/A
    Log in for availability
    Log in for availability
    Each
    Each - $574.00
    $574.00
    Each
       $574.00
    Supplier Number: 1456
  • 21511-370
    Kit, Fittings, Hitachi® Compatible
    Kit, Fittings, Hitachi® Compatible
    Multiple/Other
    Assorted
    N/A
    Discontinued
    Supplier Number: -
  • 21001-112
    Kit, Fittings
    Kit, Fittings
    ETFE, PFA
    Assorted
    N/A
    Log in for availability
    Log in for availability
    Each
    Each - $510.29
    $510.29
    Each
       $510.29
    Supplier Number: 1454
  • 53498-566
    Kit, Fittings, Hewlett-Packard® Compatible
    Kit, Fittings, Hewlett-Packard® Compatible
    Multiple/Other
    Assorted
    N/A
    Log in for availability
    Log in for availability
    Each
    Each - $528.86
    $528.86
    Each
       $528.86
    Supplier Number: 1404
  • 53500-666
    Kit, Fingertight Fittings
    Kit, Fingertight Fittings
    Multiple/Other
    Assorted
    N/A
    Discontinued
    Supplier Number: -
  • 21511-368
    Kit, Fittings, Waters® Compatible
    Kit, Fittings, Waters® Compatible
    Multiple/Other
    Assorted
    N/A
    Discontinued
    Supplier Number: -
  • 21511-372
    Kit, Fittings, Shimadzu® Compatible
    Kit, Fittings, Shimadzu® Compatible
    Multiple/Other
    Assorted
    N/A
    Discontinued
    Supplier Number: -
  • 53500-848
    Kit, Flangeless Fittings
    Kit, Flangeless Fittings
    Multiple/Other
    Assorted
    N/A
    Log in for availability
    Log in for availability
    Each
    Each - $567.99
    $567.99
    Each
       $567.99
    Supplier Number: 1451
  • 21001-114
    Kit, Fittings, High Pressure
    Kit, Fittings, High Pressure
    PEEK
    Assorted
    N/A
    Log in for availability
    Log in for availability
    Each
    Each - $1,053.86
    $1,053.86
    Each
       $1,053.86
    Supplier Number: 1457