ContactUsLinkComponent
Giant Water Prism
Giant Water Prism
Catalog # 470136-666
Supplier:  Science First
undefined
Giant Water Prism
Catalog # 470136-666
Supplier:  Science First
Supplier Number:  614-0470
Specifications
  • Description:
    Giant Water Prism
  • Environmentally Preferable:
  • Height:
    16"
  • Cat. No.:
    470136-666
  • Supplier no.:
    50-300
Specifications
About this item
Conduct Colorful Optics Experiments
  • Produce a spectrum the whole class can see by filling this transparent prism with water or mineral oil
  • Made from 1/4" thick acrylic, each panel is cut on a CNC machine and joined with the same techniques used for building aquariums
This high-quality prism is guaranteed not to leak under normal use and comes complete with spout and instructions.