ContactUsLinkComponent
VWR® Ready One-Step RT-PCR Kit
VWR® Ready One-Step RT-PCR Kit
Catalog # 71005-996
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
VWR® Ready One-Step RT-PCR Kit
Catalog # 71005-996
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ready One-Step RT-PCR Kit
  • Size:
    Kit (Sufficient material for 50 assays)
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Cat. No.:
    71005-996
  • Supplier no.:
    1B1326-50RXN
Specifications
About this item
Ready One-Step RT-PCR Kit offers a single-step procedure for performing RT-PCR reactions. Subsequent loading and visualization of RT-PCR products are streamlined since the loading buffer and visualization dye are included. The user supplies the primers and DNase-treated template RNA.
  • One-Step cDNA synthesis
  • Convenient format
  • Load directly to the gel
  • Immediate visualization after electrophoresis
  • Includes safe alternative to ethidium bromide
Ready One-Step RT-PCR Kit includes Ready One-Step Reverse Transcriptase and Ready One-Step RT-PCR Mix, 2X. Ready One-Step Reverse Transcriptase consists of M-MLV and a proprietary buffer that delivers robust cDNA synthesis from rare and abundant transcripts. Ready One-Step RT-PCR Mix, 2X consists of a mixture of reaction buffer, VWR® Extender™ Taq polymerase blend, dNTPs, an electrophoresis tracking dye, and a non-mutagenic EZ-Vision® visualization dye. Once amplification is complete, the PCR reaction can be directly loaded and separated on an agarose gel using the magenta-colored tracking dye (migrating at approximately 10bp on a 1% gel). After electrophoresis the PCR products are immediately visualized with standard UV illumination without additional post-run staining and destaining steps.