Order Entry
United States
ContactUsLinkComponent
Magnesium sulfate ≥99.0% (excluding ignited substances), powder USP, Macron Fine Chemicals™
Magnesium sulfate ≥99.0% (excluding ignited substances), powder USP, Macron Fine Chemicals™
MOC:  Regulated
Certificates
undefined
Magnesium sulfate ≥99.0% (excluding ignited substances), powder USP, Macron Fine Chemicals™
MOC:  Regulated
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Loss on Drying at 105°C:
    ≤2 %
  • Solubility:
    Passes Test
  • USP - pH (1 in 20):
    5.0 - 9.2
  • USP - Chloride (Cl):
    ≤0.014 %
  • USP - Selenium (Se):
    ≤0.003 %
  • USP - Iron (Fe):
    ≤20 ppm
  • USP - Assay (as MgSO4) (ignited basis):
    99.0 - 100.5 %
  • Thru U.S. No. 100 Sieve:
    ≥85 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together