Order Entry
United States
ContactUsLinkComponent
Sodium metabisulfite NF, Multi-Compendial
Sodium metabisulfite NF, Multi-Compendial
Catalog # JT3551-5
CAS Number:  
undefined
Sodium metabisulfite NF, Multi-Compendial
Catalog # JT3551-5
Supplier Number:  3551-05
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Poly bottle, blue T/E cap
  • Size:
    2.5 kg
  • Environmentally Preferable:
  • Cat. No.:
    JT3551-5
  • Supplier no.:
    3551-05

Specifications

Specification Test Results

  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Limit of Chloride:
    ≤ 0.05 %
  • NF - Iron (Fe):
    ≤ 0.002 %
  • NF - Limit of Thiosulfate:
    ≤ 0.05 %
  • NF - Assay (SO₂):
    65.0 - 67.4 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Thiosulfate:
    Passes Test
  • EP/BP - Iron (Fe):
    ≤ 20 ppm
  • EP/BP - Assay (Na₂S₂O₅):
    95.0 - 100.5 %
  • EP/BP - pH:
    3.5 - 5.0
  • JP - Assay (Na₂S₂O₅):
    95.0 - 100.5 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Thiosulfate:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 20 ppm
  • JP - Iron (Fe):
    ≤ 20 ppm
  • JP - Arsenic (As):
    ≤ 4 ppm

Specification Test Results

About this item

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite
 :
 :190.11 g/mol
EINECS:231-673-0
REACH:01-2119531326-45
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7681-57-4
Formula:Na₂S₂O₅
MDL Number:MFCD00167602
Melting Pt:150
Density:1.48 g/cm³ (20 °C)
Merck Index:12,08784
 :Management of Change (MOC) category = R