
di-Potassium oxalate monohydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated, With Information
About this item
CAS Number:6487-48-5
:
Formula:(COOK)₂·H₂O
MDL Number:MFCD00150033
:184.23 g/mol
Merck Index:12,07820
Melting Pt:356 °C
Density:2.154 g/cm³ (20 °C)
EINECS:209-506-8
ADR:6.1,III
:Management of Change (MOC) category = N
UN:2811
:GHS07
Storage Temperature:Ambient
Specification Test Results
- ACS - Assay (K₂C₂O₄·H₂O):98.5 - 101.0 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Neutrality:Passes Test
- ACS - pH of 5% Solution at 25°C:7.0 - 8.5
- ACS - Chloride (Cl):≤0.002 %
- ACS - Sulfate (SO₄):≤0.01 %
- ACS - Ammonium (NH₄):≤0.002 %
- ACS - Heavy Metals (as Pb):≤0.002 %
- ACS - Iron (Fe):≤0.001 %
- ACS - Sodium (Na):≤0.02 %
- ACS - Substances Darkened by Hot H₂SO₄:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Number3212 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT3212-5
Product specification
Frequently Bought Together