ContactUsLinkComponent
Slide Grip, WHEATON®, DWK Life Sciences
  14231-386
Small Business Enterprise
undefined
Slide Grip, WHEATON®, DWK Life Sciences
  14231-386
 :  900575
Small Business Enterprise
  • Description:
    Slide Grip
  • Cat. No.:
    14231-386
  • Supplier no.:
    900575
The WHEATON Slide Grip is made of polypropylene to allow for easy and safe transfer of five slides to other containers for staining
The slide grip fits into WHEATON Copling staining jars.