Order Entry
United States
ContactUsLinkComponent
 
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids
Catalog # 45000-700
Supplier:  Corning
CAS Number:  
undefined
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids
Catalog # 45000-700
Supplier:  Corning
Supplier Number:  25-025-CI
CAS Number:  

Specifications

  • Description:
    MEM, Corning® 100 ml MEM nonessential amino acids, 100X
  • Storage Temperature:
    2...8 °C
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    45000-700
  • Volume:
    100 ml
  • Supplier no.:
    25-025-CI

Specifications

About this item

The addition of MEM nonessential amino acids to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells

This concentrated supplement allows for the efficient addition to media with no risk of dilution of media components.