Order Entry
United States
ContactUsLinkComponent
Corning® HEPES buffer solution (1 mol/L)
Corning® HEPES buffer solution (1 mol/L)
Catalog # 45000-690
Supplier:  Corning
CAS Number:  
undefined
Corning® HEPES buffer solution (1 mol/L)
Catalog # 45000-690
Supplier:  Corning
Supplier Number:  25-060-CI
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Description:
    HEPES buffer solution (1 mol/l)
  • Storage Temperature:
    Ambient
  • Cat. No.:
    45000-690
  • Volume:
    100 ml
  • Supplier no.:
    25-060-CI

Specifications

About this item

HEPES (N-2-hydroxyethylpiperazine-N'-2-ethanesulphonic acid) provides an alternative to the use of bicarbonate for buffering because it does not require elevated levels of CO2

Unlike the low pKa of bicarbonate, HEPES has a pKa of 7.66 making it a much stronger buffer in the pH 7.2-7.6 range than bicarbonate buffer. Potential disadvantages of HEPES include toxicity over 100 mM and lack of nutritive value to the cell.