Order Entry
United States
ContactUsLinkComponent
Corning® HAT (Hypoxanthine, Aminopterin, Thymidine), Corning
Corning® HAT (Hypoxanthine, Aminopterin, Thymidine), Corning
Catalog # 45000-686
Supplier:  Corning
CAS Number:  
undefined
Corning® HAT (Hypoxanthine, Aminopterin, Thymidine), Corning
Catalog # 45000-686
Supplier:  Corning
Supplier Number:  25-046-CI
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Source:
  • Species:
  • Description:
    HAT (Hypoxanthine, Aminopterin, Thymidine)
  • Environmentally Preferable:
  • Storage and Stability:
    Frozen
  • Cat. No.:
    45000-686
  • Supplier no.:
    25-046-CI

Specifications

About this item

HAT Medium (hypoxanthine-aminopterin-thymidine medium) is used in mammalian cell culture for post-fusion selection to eliminate certain types of myeloma cells

The combination of aminopterin, a drug that acts as a powerful folate metabolism inhibitor, with hyproxanthine and thymidine, which are intermediates in DNA synthesis, provides a form of artificial selection for cells containing functioning hypoxanthine-guanine phosphoribosyltransferase (HGPRT) and thymidine kinase (TK). These enzymes are involved in DNA synthesis and cell division. HAT medium is often used in the preparation of monoclonal antibodies.