Corning® SSC buffer solution 20X concentrate (saline-sodium citrate)
Catalog # 45001-046
Supplier: Corning
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:SSC buffer solution 20X concentrate (saline-sodium citrate)
- Size:1 L
- Cat. No.:45001-046
- Supplier no.:46-020-CM
Specifications
About this item
20X Saline-Sodium Citrate (SSC) buffer is used in both nucleic acid hybridization and blot transfer procedures
20X is the maximum concentration used; 0.2X may be used depending on the application.