ContactUsLinkComponent
O-Ring Kit, FETFE, Ace Glass Incorporated
O-Ring Kit, FETFE, Ace Glass Incorporated
Catalog # 89235-690
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
O-Ring Kit, FETFE, Ace Glass Incorporated
Catalog # 89235-690
Supplier:  Ace Glass
Supplier Number:  7855-799
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    FETFE O-Ring Kit
  • Cat. No.:
    89235-690
  • Supplier no.:
    7855-799
Specifications
About this item
This FETFE o-ring kit is packaged in a clear, plastic box with a re-sealable lid that lets you see the o-rings inside
FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. Generally black or white in color. USP Class VI material. FETFE is a registered trademark of Ace Glass, Incorporated.
It opens to 18 compartments, each clearly marked with the o-ring size. The cardboard insert indicates the codes for each size, so you can easily re-order the o-rings individually as they are depleted. Includes 12 sizes in all, 216 total o-rings.
Ordering information: The following sizes are included: -008, -009, -010, -011, -012, -013, -014, -018, -108, -110, -111, -112, -116, -121, -136, -210, -212, and -220.